COMH4211A257 Combi-Blocks, Inc.

2-(2,4-Dimethoxyphenyl)pyridine

  • MDL number MFCD32065722
  • Cas Number : 98061-24-6
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4731

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4731
Availability details for
Quantity
  • Availability
  • Add to Cart