COMH4211A259 Combi-Blocks, Inc.

9-(Naphthalen-1-yl)-10-[4-(naphthalen-2-yl)phenyl]anthracene

  • MDL number MFCD26388421
  • Cas Number : 1026768-26-2
  • Molecular Weight : 506.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4733

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4733
Availability details for
Quantity
  • Availability
  • Add to Cart