COMH4211A25A Combi-Blocks, Inc.

2-[9-Bromo-7,7-dimethyl-13-(naphthalen-2-yl)indeno[1,2-b]anthracen-5-yl]naphthalene

  • MDL number MFCD28048411
  • Cas Number : 1196107-75-1
  • Molecular Weight : 625.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4734

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4734
Availability details for
Quantity
  • Availability
  • Add to Cart