COMH4211A25B Combi-Blocks, Inc.

9,9-Dimethyl-N-(3-phenylphenyl)fluoren-2-amine

  • MDL number MFCD30471917
  • Cas Number : 1372778-66-9
  • Molecular Weight : 361.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4735

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4735
Availability details for
Quantity
  • Availability
  • Add to Cart