COMH4211A25C Combi-Blocks, Inc.

N-(9,9-Dimethylfluoren-2-yl)-9,9-diphenylfluoren-2-amine

  • MDL number MFCD32062889
  • Cas Number : 1456702-56-9
  • Molecular Weight : 525.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4736

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4736
Availability details for
Quantity
  • Availability
  • Add to Cart