COMH4211A25E Combi-Blocks, Inc.

N-(9,9-Dimethylfluoren-2-yl)-N-(2-phenylphenyl)-9,9'-spirobi[fluorene]-7-amine

  • MDL number MFCD30543393
  • Cas Number : 1364603-07-5
  • Molecular Weight : 675.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4739

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4739
Availability details for
Quantity
  • Availability
  • Add to Cart