COMH4211A260 Combi-Blocks, Inc.

2-Bromo-11,11-dimethylbenzo[b]fluorene

  • MDL number MFCD28716947
  • Cas Number : 1198396-39-2
  • Molecular Weight : 323.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4741

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4741
Availability details for
Quantity
  • Availability
  • Add to Cart