COMH4211A261 Combi-Blocks, Inc.

2-Bromo-5,5-dimethylbenzo[b]fluorene

  • MDL number MFCD32065724
  • Cas Number : 1674334-59-8
  • Molecular Weight : 323.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4742

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4742
Availability details for
Quantity
  • Availability
  • Add to Cart