COMH4211A262 Combi-Blocks, Inc.

2'-Bromospiro[benzo[c]fluorene-7,9'-fluorene]

  • MDL number MFCD32065725
  • Cas Number : 1099593-10-8
  • Molecular Weight : 445.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4743

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4743
Availability details for
Quantity
  • Availability
  • Add to Cart