COMH4211A263 Combi-Blocks, Inc.

N-(4-Bromophenyl)-N,4-diphenylaniline

  • MDL number MFCD24387484
  • Cas Number : 503299-24-9
  • Molecular Weight : 400.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4744

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4744
Availability details for
Quantity
  • Availability
  • Add to Cart