COMH4211A264 Combi-Blocks, Inc.

2-Chloro-4-(naphthalen-2-yl)quinazoline

  • MDL number MFCD32065726
  • Cas Number : 124959-44-0
  • Molecular Weight : 290.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4745

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4745
Availability details for
Quantity
  • Availability
  • Add to Cart