COMH4211A265 Combi-Blocks, Inc.

2-Bromopyrene

  • MDL number MFCD16660390
  • Cas Number : 1714-27-8
  • Molecular Weight : 281.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4746
Availability details for
Quantity
  • Availability
  • Add to Cart