COMH4211A266 Combi-Blocks, Inc.

1,6-Dibromo-3,8-diisopropylpyrene

  • MDL number MFCD27979119
  • Cas Number : 869340-02-3
  • Molecular Weight : 444.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4747

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4747
Availability details for
Quantity
  • Availability
  • Add to Cart