COMH4211A267 Combi-Blocks, Inc.

2-(4-Bromonaphthalen-1-yl)-4,6-diphenyl-1,3,5-triazine

  • MDL number MFCD30469069
  • Cas Number : 1800228-86-7
  • Molecular Weight : 438.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4748

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4748
Availability details for
Quantity
  • Availability
  • Add to Cart