COMH4211A268 Combi-Blocks, Inc.

Methyl 3-amino-4-bromo-6-fluoro-2-methylbenzoate

  • MDL number MFCD28650861
  • Cas Number : 1784060-92-9
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4753

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4753
Availability details for
Quantity
  • Availability
  • Add to Cart