COMH4211A26B Combi-Blocks, Inc.

9-(Prop-2-yn-1-yloxy)furo[3,2-g]chromen-2-one

  • MDL number MFCD32065728
  • Cas Number : 1875054-10-6
  • Molecular Weight : 240.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4756

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4756
Availability details for
Quantity
  • Availability
  • Add to Cart