COMH4211A26C Combi-Blocks, Inc.

1-Oxo-N,2-diphenylisoquinoline-4-carboxamide

  • MDL number MFCD32065729
  • Cas Number : 2365419-39-0
  • Molecular Weight : 340.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4758

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4758
Availability details for
Quantity
  • Availability
  • Add to Cart