COMH4211A270 Combi-Blocks, Inc.

Ethyl 3-oxo-4-pentenoate

  • MDL number MFCD11041386
  • Cas Number : 22418-80-0
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8996

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-8996
Availability details for
Quantity
  • Availability
  • Add to Cart