COMH4211A271 Combi-Blocks, Inc.

1-(4-Chloro-2-methylphenyl)piperazine

  • MDL number MFCD00044555
  • Cas Number : 58820-36-3
  • Molecular Weight : 210.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4765

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4765
Availability details for
Quantity
  • Availability
  • Add to Cart