COMH4211A278 Combi-Blocks, Inc.

Benzyl quinoxaline-2-carboxylate

  • MDL number MFCD18094129
  • Cas Number : 1240842-74-3
  • Molecular Weight : 264.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 2-quinoxalinecarboxylate
Inchi Code 1S/C16H12N2O2/c19-16(20-11-12-6-2-1-3-7-12)15-10-17-13-8-4-5-9-14(13)18-15/h1-10H,11H2
Inchi Key SCEBWMVYKLIACZ-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4774

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4774
Availability details for
Quantity
  • Availability
  • Add to Cart