COMH4211A27B Combi-Blocks, Inc.

4-(3-Hydroxyphenoxy)benzonitrile

  • MDL number MFCD12763666
  • Cas Number : 1183032-44-1
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4781
Availability details for
Quantity
  • Availability
  • Add to Cart