COMH4211A27C Combi-Blocks, Inc.

Phenyl benzenesulfonate

  • MDL number MFCD00607983
  • Cas Number : 4358-63-8
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4782

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4782
Availability details for
Quantity
  • Availability
  • Add to Cart