COMH4211A27F Combi-Blocks, Inc.

Chromeno[3,4-b]pyridin-5-one

  • MDL number MFCD18451026
  • Cas Number : 65297-93-0
  • Molecular Weight : 197.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4785

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4785
Availability details for
Quantity
  • Availability
  • Add to Cart