COMH4211A281 Combi-Blocks, Inc.

2-hydroxy-3-methoxypyridine-4-carboxylic acid

  • MDL number MFCD31743283
  • Cas Number : 1935950-11-0
  • Molecular Weight : 169.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4787

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4787
Availability details for
Quantity
  • Availability
  • Add to Cart