COMH4211A284 Combi-Blocks, Inc.

1-(3-Amino-phenyl)-cyclopropanecarboxylic acid amide

  • MDL number MFCD28248789
  • Cas Number : 1456714-49-0
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9049

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9049
Availability details for
Quantity
  • Availability
  • Add to Cart