COMH4211A287 Combi-Blocks, Inc.

5-(Chloromethyl)thiophene-2-carbonitrile

  • MDL number MFCD18250243
  • Cas Number : 1260667-25-1
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4796

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4796
Availability details for
Quantity
  • Availability
  • Add to Cart