COMH4211A289 Combi-Blocks, Inc.

1-Chloro-5-methyl-4-nitro-2-(trifluoromethyl)benzene

  • MDL number MFCD04972734
  • Molecular Weight : 239.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4798

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4798
Availability details for
Quantity
  • Availability
  • Add to Cart