COMH4211A28E Combi-Blocks, Inc.

Dimethylsulfonioacetate chloride

  • MDL number MFCD20639634
  • Cas Number : 10132-50-0
  • Molecular Weight : 156.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4806

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4806
Availability details for
Quantity
  • Availability
  • Add to Cart