COMH4211A293 Combi-Blocks, Inc.

Ethyl 3-bromo-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylate

  • MDL number MFCD32065748
  • Cas Number : 2090188-35-3
  • Molecular Weight : 284.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4812

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4812
Availability details for
Quantity
  • Availability
  • Add to Cart