COMH4211A294 Combi-Blocks, Inc.

3-Bromo-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid

  • MDL number MFCD32065749
  • Cas Number : 1782049-04-0
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4813

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4813
Availability details for
Quantity
  • Availability
  • Add to Cart