COMH4211A2CC Combi-Blocks, Inc.

5-tert-Butyl-2-methylphenol

  • MDL number MFCD02667430
  • Cas Number : 5781-02-2
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4822

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4822
Availability details for
Quantity
  • Availability
  • Add to Cart