COMH4211A2CE Combi-Blocks, Inc.

Ethyl 3-(2-fluoro-5-methylphenyl)propanoate

  • MDL number MFCD18396479
  • Cas Number : 1261476-95-2
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4826

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4826
Availability details for
Quantity
  • Availability
  • Add to Cart