COMH4211A2D4 Combi-Blocks, Inc.

3-Methyl-4-(4-methylphenyl)-1H-pyrazole

  • MDL number MFCD32065751
  • Cas Number : 2047972-36-9
  • Molecular Weight : 172.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4834

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4834
Availability details for
Quantity
  • Availability
  • Add to Cart