COMH4211A2D5 Combi-Blocks, Inc.

3-Chloro-2-ethylbenzaldehyde

  • MDL number MFCD28969421
  • Cas Number : 1289030-85-8
  • Molecular Weight : 168.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4835

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4835
Availability details for
Quantity
  • Availability
  • Add to Cart