COMH4211A2D6 Combi-Blocks, Inc.

1-Bromo-3-chloro-2-ethylbenzene

  • MDL number MFCD22490780
  • Cas Number : 1780808-66-3
  • Molecular Weight : 219.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-3-chloro-2-ethylbenzene
Inchi Code 1S/C8H8BrCl/c1-2-6-7(9)4-3-5-8(6)10/h3-5H,2H2,1H3
Inchi Key RHQXANFHLBLUCK-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4836

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4836
Availability details for
Quantity
  • Availability
  • Add to Cart