COMH4211A2D8 Combi-Blocks, Inc.

1-Fluoro-4-phenyl-2-(trifluoromethyl)benzene

  • MDL number MFCD08457036
  • Cas Number : 1032506-93-6
  • Molecular Weight : 240.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4838

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4838
Availability details for
Quantity
  • Availability
  • Add to Cart