COMH4211A2DA Combi-Blocks, Inc.

Methyl 2,6-dibromo-3-fluorobenzoate

  • MDL number MFCD28784667
  • Cas Number : 1804417-05-7
  • Molecular Weight : 311.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4840

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4840
Availability details for
Quantity
  • Availability
  • Add to Cart