COMH4211A2DB Combi-Blocks, Inc.

Methyl 2-[6-(trifluoromethyl)pyridin-2-yl]acetate

  • MDL number MFCD28964195
  • Cas Number : 1346534-09-5
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4841

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4841
Availability details for
Quantity
  • Availability
  • Add to Cart