COMH4211A2DD Combi-Blocks, Inc.

3-Bromo-5-ethylphenol

  • MDL number MFCD16997630
  • Cas Number : 749930-37-8
  • Molecular Weight : 201.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4843

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4843
Availability details for
Quantity
  • Availability
  • Add to Cart