COMH4211A2DF Combi-Blocks, Inc.

Methyl 3-oxo-4H-quinoxaline-6-carboxylate

  • MDL number MFCD24564549
  • Cas Number : 357637-38-8
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4847

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4847
Availability details for
Quantity
  • Availability
  • Add to Cart