COMH4211A2E2 Combi-Blocks, Inc.

tert-Butyl 4-{[(2-methoxyethyl)carbamoyl]methyl}piperazine-1-carboxylate

  • MDL number MFCD32065752
  • Cas Number : 2365419-48-1
  • Molecular Weight : 301.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4850

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4850
Availability details for
Quantity
  • Availability
  • Add to Cart