COMH4211A2E5 Combi-Blocks, Inc.

Ethyl 4-((tert-butoxycarbonyl)amino)-1H-pyrrole-2-carboxylate

  • MDL number MFCD24445026
  • Cas Number : 903570-20-7
  • Molecular Weight : 254.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4856

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4856
Availability details for
Quantity
  • Availability
  • Add to Cart