COMH4211A2E6 Combi-Blocks, Inc.

1-Chloro-3-(3-chloropropyl)benzene

  • MDL number MFCD09744260
  • Cas Number : 90347-05-0
  • Molecular Weight : 189.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4857

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4857
Availability details for
Quantity
  • Availability
  • Add to Cart