COMH4211A2E9 Combi-Blocks, Inc.

Mal-peg4-ala-ala-asn-pab

  • MDL number MFCD30182035
  • Cas Number : 2055042-63-0
  • Molecular Weight : 949.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9071

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-9071
Availability details for
Quantity
  • Availability
  • Add to Cart