COMH4211A2EA Combi-Blocks, Inc.

tert-Butyl 2-chloro-6-methoxypyridine-3-carboxylate

  • MDL number MFCD30189383
  • Cas Number : 2089378-14-1
  • Molecular Weight : 243.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4861

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4861
Availability details for
Quantity
  • Availability
  • Add to Cart