COMH4211A2EC Combi-Blocks, Inc.

Ethyl 4-(3,5-dimethylphenyl)butanoate

  • MDL number MFCD32065754
  • Cas Number : 73896-10-3
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4864

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4864
Availability details for
Quantity
  • Availability
  • Add to Cart