COMH4211A2F4 Combi-Blocks, Inc.

[4-(methanesulfonylmethyl)phenyl]methanol

  • MDL number MFCD26704548
  • Cas Number : 1227370-65-1
  • Molecular Weight : 200.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4881

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4881
Availability details for
Quantity
  • Availability
  • Add to Cart