COMH4211A2F9 Combi-Blocks, Inc.

1,3-Diethyl 2-acetamido-2-[(2-nitrophenyl)methyl]propanedioate

  • MDL number MFCD00483638
  • Cas Number : 6960-47-0
  • Molecular Weight : 352.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4888

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4888
Availability details for
Quantity
  • Availability
  • Add to Cart