COMH4211A2FD Combi-Blocks, Inc.

6-Methanesulfonylpyridine-2-carboxamide

  • MDL number MFCD32065760
  • Cas Number : 2365418-48-8
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4892

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4892
Availability details for
Quantity
  • Availability
  • Add to Cart