COMH4211A2FE Combi-Blocks, Inc.

Methyl 1-{[4-(methylsulfanyl)benzene]sulfonyl}piperidine-4-carboxylate

  • MDL number MFCD09677953
  • Cas Number : 928938-17-4
  • Molecular Weight : 329.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4893

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-4893
Availability details for
Quantity
  • Availability
  • Add to Cart